ENA387772807 Enamine

6-cyclobutylpyrimidin-4-ol

  • MDL number MFCD22394139
  • Cas Number : 1423024-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120065
Availability details for
Quantity
  • Availability
  • Add to Cart