ENA387772810 Enamine

(2S)-2-[(methoxycarbonyl)amino]propanoic acid

  • MDL number MFCD12795173
  • Cas Number : 59190-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120068
Availability details for
Quantity
  • Availability
  • Add to Cart