ENA387772811 Enamine

5-ethyl-1-methyl-1H-pyrazol-4-amine

  • MDL number MFCD08700618
  • Cas Number : 1007541-15-2
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120069
Availability details for
Quantity
  • Availability
  • Add to Cart