ENA387772821 Enamine

5-amino-2-(2,2,2-trifluoroethoxy)benzamide

  • MDL number MFCD14621851
  • Cas Number : 1247057-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120088
Availability details for
Quantity
  • Availability
  • Add to Cart