ENA387772823 Enamine

2-amino-4-ethoxythiophene-3-carbonitrile

  • MDL number MFCD20702489
  • Cas Number : 105558-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118763
Availability details for
Quantity
  • Availability
  • Add to Cart