ENA387772824 Enamine

1-{[1-(bromomethyl)cyclobutyl]methoxy}-3-(methoxymethyl)benzene

  • MDL number MFCD20886095
  • Cas Number : 1423025-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120109
Availability details for
Quantity
  • Availability
  • Add to Cart