ENA387772826 Enamine

2-methyl-6-phenoxyquinolin-4-ol

  • MDL number MFCD03847529
  • Cas Number : 56716-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120121
Availability details for
Quantity
  • Availability
  • Add to Cart