ENA387772827 Enamine

2-(1-ethyl-1H-pyrazol-4-yl)-2,2-difluoroacetic acid

  • MDL number MFCD16748881
  • Cas Number : 1249324-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120128
Availability details for
Quantity
  • Availability
  • Add to Cart