ENA387772828 Enamine

tert-butyl 3-(3,4-difluorophenyl)-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD22628495
  • Cas Number : 1227617-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120129
Availability details for
Quantity
  • Availability
  • Add to Cart