ENA387772829 Enamine

2-[2-(benzyloxy)-4-methoxyphenyl]propan-2-ol

  • MDL number MFCD22628496
  • Cas Number : 1423032-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120130

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120130
Availability details for
Quantity
  • Availability
  • Add to Cart