ENA387772832 Enamine

(2-cyclobutoxypyridin-4-yl)methanamine

  • MDL number MFCD16684252
  • Cas Number : 1250643-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120134
Availability details for
Quantity
  • Availability
  • Add to Cart