ENA387772834 Enamine

4-cyclopropyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD19282048
  • Cas Number : 1266822-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120143

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120143
Availability details for
Quantity
  • Availability
  • Add to Cart