ENA387772835 Enamine

3-(4-chlorophenyl)cyclopent-2-en-1-ol

  • MDL number MFCD21129877
  • Cas Number : 1423025-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120144
Availability details for
Quantity
  • Availability
  • Add to Cart