ENA387772839 Enamine

4-tert-butylpiperidine-1-sulfonyl chloride

  • MDL number MFCD16671558
  • Cas Number : 1251076-14-8
  • Molecular Weight : 239.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120148
Availability details for
Quantity
  • Availability
  • Add to Cart