ENA387772841 Enamine

cyclobutanecarboximidamide hydrochloride

  • MDL number MFCD09757543
  • Cas Number : 71690-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120150

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120150
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart