ENA387772845 Enamine

2-methyl-5-phenylpyridin-3-amine

  • MDL number MFCD18260244
  • Cas Number : 1256834-25-9
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-5-phenyl-3-pyridinamine
Inchi Code 1S/C12H12N2/c1-9-12(13)7-11(8-14-9)10-5-3-2-4-6-10/h2-8H,13H2,1H3
Inchi Key VZAGXOBOKSVVJK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120155
Availability details for
Quantity
  • Availability
  • Add to Cart