ENA387772852 Enamine

(1R)-1-(3-chlorophenyl)propan-1-ol

  • MDL number MFCD18339565
  • Cas Number : 112777-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120163
Availability details for
Quantity
  • Availability
  • Add to Cart