ENA387772853 Enamine

2-(oxolan-2-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD19264298
  • Cas Number : 1266936-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118838
Availability details for
Quantity
  • Availability
  • Add to Cart