ENA387772855 Enamine

(1S)-1-(3-chlorophenyl)propan-1-ol

  • MDL number MFCD18339532
  • Cas Number : 172748-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120167
Availability details for
Quantity
  • Availability
  • Add to Cart