ENA387772859 Enamine

2-{[(tert-butoxy)carbonyl]amino}-4-(dimethylamino)butanoic acid

  • MDL number MFCD19401087
  • Cas Number : 1404828-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120171
Availability details for
Quantity
  • Availability
  • Add to Cart