ENA387772862 Enamine

2-[(2-methoxyphenyl)methyl]cyclopentan-1-one

  • MDL number MFCD16073835
  • Cas Number : 2866-63-9
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120175
Availability details for
Quantity
  • Availability
  • Add to Cart