ENA387772870 Enamine

4-[methyl(phenyl)carbamoyl]benzoic acid

  • MDL number MFCD12466761
  • Cas Number : 153849-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118914
Availability details for
Quantity
  • Availability
  • Add to Cart