ENA387772872 Enamine

2,3-dichloro-4-methylaniline

  • MDL number MFCD20690792
  • Cas Number : 80026-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120238
Availability details for
Quantity
  • Availability
  • Add to Cart