ENA387772873 Enamine

4-(5-chloro-1H-indol-1-yl)butanoic acid

  • MDL number MFCD18900021
  • Cas Number : 1229953-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120239
Availability details for
Quantity
  • Availability
  • Add to Cart