ENA387772875 Enamine

ethyl 7-chloro-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate

  • MDL number MFCD20922945
  • Cas Number : 16867-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120241
Availability details for
Quantity
  • Availability
  • Add to Cart