ENA387772876 Enamine

5-amino-1-(propan-2-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD03424118
  • Cas Number : 21254-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120244

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120244
Availability details for
Quantity
  • Availability
  • Add to Cart