ENA387772890 Enamine

3-(3-methyl-4H-1,2,4-triazol-4-yl)aniline

  • MDL number MFCD21984471
  • Cas Number : 361549-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120262

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120262
Availability details for
Quantity
  • Availability
  • Add to Cart