ENA387772901 Enamine

4-(1H-1,2,3-triazol-1-yl)phenol

  • MDL number MFCD22628513
  • Cas Number : 68535-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120293
Availability details for
Quantity
  • Availability
  • Add to Cart