ENA387772908 Enamine

methyl 4-hydrazinyl-3-methoxybenzoate

  • MDL number MFCD19201437
  • Cas Number : 952285-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118994

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118994
Availability details for
Quantity
  • Availability
  • Add to Cart