ENA387772912 Enamine

quinolin-8-ylmethanesulfonamide

  • MDL number MFCD11212982
  • Cas Number : 1094691-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120349

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120349
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart