ENA387772917 Enamine

5-(4-methyl-4H-1,2,4-triazol-3-yl)pyridin-2-amine

  • MDL number MFCD18304450
  • Cas Number : 1339183-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120356
Availability details for
Quantity
  • Availability
  • Add to Cart