ENA387772929 Enamine

2-(4-chlorophenyl)-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD00220089
  • Cas Number : 174422-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 319...321
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120386
Availability details for
Quantity
  • Availability
  • Add to Cart