ENA387772934 Enamine

2-(2,5-dichlorophenyl)pyrrolidine

  • MDL number MFCD02663510
  • Cas Number : 383127-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120393
Availability details for
Quantity
  • Availability
  • Add to Cart