ENA387772944 Enamine

4-(trifluoromethyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD11035860
  • Cas Number : 266359-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119065
Availability details for
Quantity
  • Availability
  • Add to Cart