ENA387772962 Enamine

4-ethylpyrrolidin-3-ol

  • MDL number MFCD22628533
  • Cas Number : 1423027-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120462
Availability details for
Quantity
  • Availability
  • Add to Cart