ENA387772969 Enamine

2-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-1,3-benzodiazole

  • MDL number MFCD22628536
  • Cas Number : 26663-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120470
Availability details for
Quantity
  • Availability
  • Add to Cart