ENA387772971 Enamine

4-amino-N-(1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide

  • MDL number MFCD00592980
  • Cas Number : 16806-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120472
Availability details for
Quantity
  • Availability
  • Add to Cart