ENA387772973 Enamine

1-(3-chloropyridin-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD14594767
  • Cas Number : 1250109-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120475
Availability details for
Quantity
  • Availability
  • Add to Cart