ENA387772974 Enamine

4,8-dimethylnona-1,3,7-triene

  • MDL number MFCD13180489
  • Cas Number : 51911-82-1
  • Molecular Weight : 150.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3E)-4,8-dimethyl-1,3,7-nonatriene
Inchi Code 1S/C11H18/c1-5-7-11(4)9-6-8-10(2)3/h5,7-8H,1,6,9H2,2-4H3/b11-7+
Inchi Key LUKZREJJLWEWQM-YRNVUSSQSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120476

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P273,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-120476
pictograms GHS02,GHS07
hstatements H226,H315,H319,H335,H412
Availability details for
Quantity
  • Availability
  • Add to Cart