ENA387772975 Enamine

4-bromo-1H-indole-6-carbonitrile

  • MDL number MFCD03095209
  • Cas Number : 374633-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120478
Availability details for
Quantity
  • Availability
  • Add to Cart