ENA387772977 Enamine

1H-imidazo[4,5-c]pyridin-2-amine

  • MDL number MFCD02237634
  • Cas Number : 68074-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120481
Availability details for
Quantity
  • Availability
  • Add to Cart