ENA387772984 Enamine

4-(2,4-difluorophenyl)oxan-4-amine

  • MDL number MFCD18296266
  • Cas Number : 1343080-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120519
Availability details for
Quantity
  • Availability
  • Add to Cart