ENA387772986 Enamine

7-methyl-1H,2H,3H-pyrido[2,3-b][1,4]oxazine

  • MDL number MFCD18260627
  • Cas Number : 1198154-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120522
Availability details for
Quantity
  • Availability
  • Add to Cart