ENA387772987 Enamine

5,5-dimethyl-2,3,4,5-tetrahydro-1H-1-benzazepine

  • MDL number MFCD18375171
  • Cas Number : 22991-42-0
  • Molecular Weight : 175.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,5-dimethyl-2,3,4,5-tetrahydro-1H-1-benzazepine
Inchi Code 1S/C12H17N/c1-12(2)8-5-9-13-11-7-4-3-6-10(11)12/h3-4,6-7,13H,5,8-9H2,1-2H3
Inchi Key NRSCJWAKHJNQEP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120523
Availability details for
Quantity
  • Availability
  • Add to Cart