ENA387772994 Enamine

4-[(4-hydroxycyclohexyl)amino]-3-methylbenzonitrile

  • MDL number MFCD12872523
  • Cas Number : 1178262-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119074

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119074
Availability details for
Quantity
  • Availability
  • Add to Cart