ENA387772996 Enamine

4-(4-acetylphenoxy)benzoic acid

  • MDL number MFCD00570558
  • Cas Number : 1274135-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120532
Availability details for
Quantity
  • Availability
  • Add to Cart