ENA387773000 Enamine

2-amino-3,4,6-trichlorophenol

  • MDL number MFCD06252434
  • Cas Number : 6358-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119075
Availability details for
Quantity
  • Availability
  • Add to Cart