ENA387773006 Enamine

1-(1,3-thiazol-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD14594687
  • Cas Number : 1250077-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119076
Availability details for
Quantity
  • Availability
  • Add to Cart