ENA387773014 Enamine

ethyl 2-benzoylbutanoate

  • MDL number MFCD16077033
  • Cas Number : 24346-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120583
Availability details for
Quantity
  • Availability
  • Add to Cart