ENA387773018 Enamine

4,6-dichloro-N-cyclopropyl-1,3,5-triazin-2-amine

  • MDL number MFCD10566402
  • Cas Number : 32889-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120590
Availability details for
Quantity
  • Availability
  • Add to Cart